Magnesium chloride 1 M in aqueous solution, OmniPur®, sterile, Millipore®

5985
EM-5985EA 128.04 USD
EM-5985
Magnesium chloride 1 M in aqueous solution, OmniPur®, sterile, Millipore®
Magnesium chloride

Sterile solution ideal for preparation of competent cells for transfection. Molecular biology grade.


  • DNase-, RNase-, and protease-free
  • Clear, colorless liquid


A solution of 9.5% magnesium chloride and 90.5% water.

Formula: MgCl₂
MW: 95.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR