Mercury (II) oxide ≥99.998% (metals basis), Puratronic®

Danger

10810-14 10810-14
AA10810-14EA 253.32 USD
CAAA10810-14 AA10810-14
Mercury (II) oxide ≥99.998% (metals basis), Puratronic®
Mercury (II) oxide

Powder. Purity is based on metal contaminates only.

Formula: HgO
MW: 216.59 g/mol
Melting Pt: 500 °C (dec.)
Density: 11.14
Storage Temperature: Ambient
MDL Number: MFCD00011045
CAS Number: 21908-53-2
EINECS: 244-654-7
UN: 1641
ADR: 6.1,II
Merck Index: 13,05908

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR