Lanthanum, 1% w/v , J.T.Baker®
BAKR6948-01 BAKR6948-01
JT6948-1CS 863.75 USD
JT6948-1 CAJT6948-1
Lanthanum, 1% w/v , J.T.Baker®
Spectral releasing agent. Lanthanum nitrate 99.999% spectral purity in 0.3M nitric acid. The metal content of the solution confirmed by titration.