Lithium carbonate ≥99.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning


Management of Change (MOC) category = N

BAKR2362-01 BAKR2362-01
JT2362-1CS 1306.46 USD
JT2362-1 CAJT2362-1
Lithium carbonate ≥99.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Lithium carbonate

Powder. Lot analysis on label.

Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720 °C
Density: 2.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Li₂CO₃) (by acidimetry) ≥99.0 %
Insoluble in Dilute HCl ≤0.01 %
Chloride (Cl) ≤0.005 %
Total Sulfur (as SO₄) ≤0.2 %
Heavy Metals (as Pb) ≤0.002 %
Iron (Fe) ≤0.002 %
Calcium (Ca) ≤0.01 %
Potassium (K) ≤0.005 %
Silicon (Si), For Information Only
Sodium (Na) ≤0.010 %
Nitrate (NO₃) ≤5 ppm
Specific Surface Area, m²/g, For Information Only
Bulk Density (g/cc), For Information Only
Mesh (Wet Screen Analysis) - On U.S. No. 325 Sieve, For Information Only

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR