Iodine 1.0 N, StandARd® volumetric solution, Macron Fine Chemicals™

Danger


Management of Change (MOC) category = N

MACRH185-05 MACRH185-05
MKH18505CS 2143.46 USD
CAMKH18505 MKH18505
Iodine 1.0 N, StandARd® volumetric solution, Macron Fine Chemicals™
Iodine

Contains 10 to 20% iodine and 20 to 30% potassium iodide.

Formula: I₂
MW: 253.81 g/mol
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 3495
ADR: 8, 6.1,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Normality 0.95 - 1.05

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR