Iron(III) sulfate hydrate ≥73%, BAKER ANALYZED®, J.T.Baker®

Warning

Synonyms: Ferric sulfate hydrateDiiron trissulfate hydrateDiiron trissulphate hydrateFerric sulphate hydrate

Management of Change (MOC) category = N

BAKR2046-05 2046-05
JT2046-5CS 4667.09 USD
JT2046-5 CAJT2046-5
Iron(III) sulfate hydrate ≥73%, BAKER ANALYZED®, J.T.Baker®
Iron (III) sulphate hydrate
Formula: Fe₂(SO₄)₃·xH₂O
MW: 399.88 g/mol (anhy)
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149714
CAS Number: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (Fe₂(SO₄)₃) ≥73.0 %
Insoluble Matter ≤0.02 %
Chloride (Cl) ≤0.002 %
Nitrate (NO₃) ≤0.01 %
Copper (Cu) ≤0.005 %
Ferrous Iron ≤0.02 %
Substances Not Precipitated by NH₄OH (as SO₄) ≤0.1 %
Zinc (Zn) ≤0.005 %
Appearance (yellowish to brown powder)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR