VWR® Filtering Flasks with Capacity Scale, Glass
VW27060-4000 VW27060-250 VW27060-2000 VW27060-25 VW27060-125 VW27060-1000 VW27060-500 VW27060-50
89000-391CS 183.86 USD
89000-391 89000-382 89000-384 89005-196 89000-386 89000-376 89000-389 89000-378
VWR® Filtering Flasks with Capacity Scale, Glass
Flasks feature a heavier wall than standard Erlenmeyer flasks, and withstand vacuum to 1atm or 760 mm Hg (29in. Hg).
  • Available in a variety of sizes
Side hose connection on 25 mL through 500 mL flasks accepts 7.9mm (5/16") I.D
Meet ASTM Specification E1406, Type II, Class I requirements.
flexible tubing; sizes 1000 mL and larger accept 9.5mm (3/8") I.D. tubing.