Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PYREXPLUS® Plastic-Coated Filtering Flasks, Graduated, Heavy Wall, Corning®

Supplier: Corning
PYREXPLUS®
65340-250 65340-4L 65340-2L 65340-500 65340-1L
PYREXPLUS® labware allows for a better hand grip and clamp hold

Caution: Do not place plastic-coated ware over direct heat or flames.

PVC coating makes cleanup faster and easier by reducing spill volume if breakage occurs. Coating is autoclavable to 121°C (250°F). Do not heat above 121°C (250°F) using moist heat or 110°C (230°F) using dry heat. Resistant to thermal shock. Flasks are graduated to show approximate volume. To allow easy connection of standard size tubing, side arm is not coated.



 

ORDER

Expand All / Collapse All
Capacity Tubulation O.D. Rubber Stopper Size Supplier No. VWR Catalog Number Unit Price Quantity
1 L 10 mm 8 65340-1L 89091-676 Case of 6 Retrieving
250 mL 10 mm 6 65340-250 29410-988 Case of 6 Retrieving
2 L 12.5 mm 9 65340-2L 89090-584 Case of 4 Retrieving
Pack of 1 Retrieving
4 L 12.5 mm 12 65340-4L 89090-588 Case of 2 Retrieving
Pack of 1 Retrieving
500 mL 10 mm 7 65340-500 89091-694 Case of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty