n-Hexacosane ≥99.0%

Supplier: TCI America

Synonyms: N-hexacosane

H0050-5G H0050-25G
TCH0050-025GEA 235.52 USD
TCH0050-025G TCH0050-005G
n-Hexacosane ≥99.0%
n-Hexacosane

CAS Number: 630-01-3
MDL Number: MFCD00009354
Molecular Formula: C26H54
Molecular Weight: 366.72
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 268
Melting point (°C): 57
Flash Point (°C): 166

Formula: H₃C(CH₂)₂₄CH₃
MW: 366.71 g/mol
Boiling Pt: 268 °C
Melting Pt: 57 °C
Density: 0.804 g/cm³ (20 °C)
Flash Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00009354
CAS Number: 630-01-3
EINECS: 211-124-1
https://us.vwr.com/stibo/web/std.lang.all/01/26/28970126.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR