Hydrochloric acid 10%, StandARd®, Macron Fine Chemicals™

Danger

Synonyms: Muriatic acid

Management of Change (MOC) category = NI

H151-07 MACRH151-07
MKH15107CS 442.27 USD
CAMKH15107 MKH15107
Hydrochloric acid 10%, StandARd®, Macron Fine Chemicals™
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay ((HCl))(g/100 mL) 9.5 - 10.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR