Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PYREX® Shaker Erlenmeyer Flasks, Corning®

Supplier: Corning
PYREX®
4442-500 4442-125 1500-125 4442-2L 4442-1L
These Erlenmeyer flasks have a long, straight rimless neck of heavy tubing to reduce splashing, making them ideal for shaker cultures

Uniform wall thickness provides balance between mechanical strength and thermal shock resistance. Flasks feature marking spot for easy labeling and identification.



 

ORDER

Expand All / Collapse All
Description Capacity Stopper No. O.D. x H Neck Diameter Supplier No. VWR Catalog Number Unit Price Quantity
- 500 mL 101 x 181 mm (4 x 71/8") 38 mm (11/2") - 4442-500 29174-150 Case of 1 Retrieving
- 125 mL 66 x 122 mm (23/5 x 44/5") 25 mm (1") - 4442-125 89090-645 Case of 12 Retrieving
Pack of 6 Retrieving
- 1 L 129 x 216 mm (51/16 x 81/2") 38 mm (11/2") - 4442-1L 29174-152 Case of 1 Retrieving
- 2 L 160 x 285 mm (65/16 x 111/4") 38 mm (11/2") - 4442-2L 32645-014 Case of 1 Retrieving
Product available on GSA Advantage®
PYREX® Reagent Bottle Narrow Mouth with Stopper 125 mL (4.2 oz.) - - 19 1500-125 89090-652 Case of 48 Retrieving
Pack of 6 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty