Hematoxylin, BAKER ANALYZED®, Certified Stain for histology, for cytology, J.T.Baker®
Warning
|
Synonyms:
(6aS,11bR)-6,6a,7,11b-Tetrahydroindeno[2,1-c]chromene-3,4,6a,9,10-pentaol
, Hydroxybrasilin
, NSC 2785
, DcH-48
, Hydroxybrazilin
, (+)-Hematoxylin
, Natural Black I
Management of Change (MOC) category = NI
M906-03
BAKRM906-03
JTM906-3CS
1005.17
USD
JTM906-3
CAJTM906-3
Hematoxylin, BAKER ANALYZED®, Certified Stain for histology, for cytology, J.T.Baker®
Hematoxylin
Certified for Use in Histology and Cytology (C.I. 75290)
Formula:
C₁₆H₁₄O₆ MW: 302.28 g/mol Melting Pt: 291…293 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 EINECS: 208-237-3 Merck Index: 13,04654 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Biological Test | Passes Test |
Certified by the Biological Stain Commission | |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...