TBE Buffer, Concentrate, OmniPur*. 5X.
8800 8800-4L
EM-8800EA 128.39 USD
EM-8800 CA8800
TBE Buffer, Concentrate, OmniPur*. 5X.
Liquid
Each liter of concentrate makes 5L of 1X TBE (1X=0.089M TRIS, 0.089M Borate, 0.002M EDTA). For molecular biology. DNase-, RNase-, and protease-free.