Hydroxylammonium chloride ≥96.0% (by titrimetric analysis), crystals, BAKER ANALYZED® ACS, J.T.Baker®

Danger

Synonyms: Hydroxylamine hydrochlorideHydroxylamine HCl

Management of Change (MOC) category = N

BAKR2195-04 BAKR2195-01
JT2195-1CS 10280.25 USD
JT2195-1 JT2195-4
Hydroxylammonium chloride ≥96.0% (by titrimetric analysis), crystals, BAKER ANALYZED® ACS, J.T.Baker®
Hydroxylammonium chloride
Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8 (6.1),III
Merck Index: 13,04853

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (NH₂OH·HCl) (by KMnO₄ titrn) ≥96.0 %
Clarity of Alcohol Solution Passes Test
Residue after Ignition ≤0.050 %
Titrable Free Acid (meq/g) ≤0.25
Ammonium (NH₄) ≤0.1 %
Sulfur Compounds (as SO₄) ≤0.005 %
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm
Appearance (fine, white crystalline powder)
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR