Hydroxylammonium chloride, GR ACS, Supelco®

Warning

Synonyms: Hydroxylamine hydrochlorideHydroxylamine HCl

HX0770-1 HX0770-6
CAHX0770-1 CAHX0770-6
Hydroxylammonium chloride, GR ACS, Supelco®
Hydroxylammonium chloride

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: NH₂OH·HCl
MW: 69.49 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 151 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051089
CAS Number: 5470-11-1
EINECS: 226-798-2
UN: 2923
ADR: 8,III
Merck Index: 13,04853

Order Now

Specification Test Results

Assay 96.0% min
Ammonium (NH4) 0.1% max
Clarity of alcohol solution To pass test
Color White
Form Crystals
Heavy metals (as Pb) 5ppm max
Iron (Fe) 5ppm max
Residue after ignition 0.05% max
Sulfur compounds (as SO4) 0.005% max
Titrable free acid 0.25meq/g max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR