PYREX® Micro Distilling Flasks, Corning®
4963A-50 4963A-25
29280-769CS 181.43 USD
29280-769 29280-780
PYREX® Micro Distilling Flasks, Corning®
Flasks have three 14/20 [ST] joint necks at same height
Center neck is vertical and two side necks are angled.