Hydrofluoric acid solution 48.5 - 50.5% ACS, Supelco®
|
Danger
|
HX0621-2
HX0621-3
HX0621-2
EM-HX0621-3CS
3611.24
USD
CAHX0621-2
EM-HX0621-3
EM-HX0621-2
Hydrofluoric acid solution 48.5 - 50.5% ACS, Supelco®
Hydrofluoric acid
Formula:
HF MW: 20.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011346 CAS Number: 7664-39-3 EINECS: 231-634-8 UN: 1790 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...