HEPES sodium salt, OmniPur®, Millipore®

Synonyms: N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium saltSodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonateSodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonateSodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonateSodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulfonate4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium saltN-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt

5380-100GM 5390-1KG 5380-100GM 5390-1KG 5400-5KG
EM-5400EA 4185.09 USD
CA5390 EM-5400 EM-5390 EM-5380 CA-EM5380
HEPES sodium salt, OmniPur®, Millipore®
HEPES sodium salt

For molecular biology. DNase-, RNase-, protease-, and phosphatase-free.

Formula: C₈H₁₇N₂NaO₄S
MW: 260.29 g/mol
Melting Pt: ? 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS Number: 75277-39-3
EINECS: 278-169-7
https://us.vwr.com/stibo/web/std.lang.all/61/70/18366170.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR