Glycidyl methyl ether ≥85% (by GC)
|
Danger
|
Synonyms:
1,2-Epoxy-3-methoxypropane
G0203-25ML
TCG0203-025MLEA
105
USD
TCG0203-025ML
Glycidyl methyl ether ≥85% (by GC)
Glycidyl methyl ether
CAS Number: 930-37-0
MDL Number: MFCD00005140
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 25
Specific Gravity (20/20): 0.99
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,2-Epoxy-3-methoxypropane
Formula:
C4H8O2 Boiling Pt: 110 °C Density: 0.99 (20 °C) Flash Pt: 25 °C |
MDL Number:
MFCD00005140 CAS Number: 930-37-0 UN: 1992 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...