Glycolic acid ≥98%, J.T. Baker®

Supplier: Avantor
Danger

Synonyms: 2-Hydroxy acid

M821-05 M821-05
JTM821-5EA 152.9 USD InStock
CAJTM821-5 JTM821-5
Glycolic acid ≥98%, J.T. Baker®
Glycolic acid

Crystals.

Formula: C₂H₄O₃
MW: 76.05 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 79…80 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3261
ADR: 8,II
Merck Index: 13,04511

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₂H₄O₃) (by acid-base titration) ≥ 98 %
Appearance Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR