Fumaric acid di-(2-ethylhexyl) ester ≥98% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
Di(2-ethylhexyl) Fumarate
, Bis(2-ethylhexyl) Fumarate
, Dioctyl Fumarate
, Fumaric acid di(2-ethylhexyl) ester
, Fumaric acid dioctyl ester
CAS Number: 141-02-6
MDL Number: MFCD00065144
Molecular Formula: C20H36O4
Molecular Weight: 340.50
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 190
Specific Gravity (20/20): 0.94
Product Categories: Materials Chemistry, Resin Additives, Copolymerization Monomers
Other Product Names / Synonyms: Fumaric Acid Di(2-ethylhexyl) Ester, Di(2-ethylhexyl) Fumarate, Dioctyl Fumarate, Fumaric Acid Dioctyl Ester
Formula:
C20H36O4 Boiling Pt: 185 °C Density: 0.94 (20 °C) Flash Pt: 190 °C |
MDL Number:
MFCD00065144 CAS Number: 141-02-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...