Formic acid ≥88.0%, AR® ACS, Macron Fine Chemicals™

Danger

Synonyms: Methanoic acid

Management of Change (MOC) category = N

MACR2592-05 MACR2592-45 MACR2592-45 MACR2592-18 MACR2592-18
MK259218CS 1397.27 USD
MK259218 CAMK259245 MK259205 MK259245 CAJT2592-18
Formic acid ≥88.0%, AR® ACS, Macron Fine Chemicals™
Formic acid
Formula: HCO₂H
MW: 46.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3),II
Merck Index: 13,04265
https://us.vwr.com/stibo/web/std.lang.all/37/23/723723.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HCOOH) ≥88.0 %
ACS - Acetic Acid (CH₃COOH) ≤0.4 %
ACS - Ammonium (NH₄) ≤0.005 %
ACS - Chloride (Cl) ≤0.001 %
ACS - Color (APHA) ≤15
Dilution Test Passes Test
ACS - Heavy Metals (as Pb) ≤0.0005 %
ACS - Iron (Fe) ≤0.0005 %
ACS - Residue after Evaporation ≤0.002 %
ACS - Sulfate (SO₄) ≤0.002 %
ACS - Sulfite (SO₃) Passes Test
Appearance (clear, colorless liquid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR