Water with 0.1% (v/v) Formic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™

LC452-1 LC452-2.5
CABJLC452-2.5 CABJLC452-1
Water with 0.1% (v/v) Formic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Water with 0.1% (v/v) Formic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk


Available in 2 levels of purity: LabReady™ HPLC & LabReady™ LC-MS.

Density: 0.997 g/cm3
Storage Temperature: Ambient
EINECS: 231-791-2
REACH: 01-2119491174-37-XXXX

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR