N-Ethylacetamide ≥99.0%

Supplier: TCI America

E0324-25ML
TCE0324-025MLEA 70.01 USD
TCE0324-025ML
N-Ethylacetamide ≥99.0%
N-Ethylacetamide

CAS Number: 625-50-3
MDL Number: MFCD00009029
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 206
Melting point (°C): -32
Flash Point (°C): 110
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: N-Acetylethylamine

Formula: C₄H₉NO
Boiling Pt: 206 °C
Melting Pt: –32 °C
Density: 0.92 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009029
CAS Number: 625-50-3
EINECS: 210-896-7
https://us.vwr.com/stibo/web/std.lang.all/78/56/28967856.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR