Eosin Y (yellowish), BAKER ANALYZED®, Certified Stain for histology, J.T.Baker®

Warning

Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

Management of Change (MOC) category = NI

BAKRL088-03 L088-03
JTL088-3CS 430.1 USD
JTL088-3 CAJTL088-3
Eosin Y (yellowish), BAKER ANALYZED®, Certified Stain for histology, J.T.Baker®
Eosin Y (yellowish)

Certified for histology and compounding of blood stains. Lot analysis on label.

Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Melting Pt: 295…296 °C
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
EINECS: 241-409-6
Merck Index: 13,03634

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Total Dye Content, For Information Only
Absorbance Maximum, nm, For Information Only
Absorbance at Maximum (1.0 mg/200 mL in 0.01% Na₂CO₃,1-cm path), For Information Only
Biological Test Passes Test
Certified by the Biological Stain Commission

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR