Diethyl ether ≥98% (by GC) stabilized, BAKER ANALYZED® ACS, J.T.Baker®

Danger

Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-OxydiethaneEthyl ether

Management of Change (MOC) category = N

BAKR9240-07 BAKR9240-07 BAKR9240-05 BAKR9240-R BAKR9240-22 BAKR9240-03 BAKR9240-03 BAKR9240-R BAKR9240-05 BAKR9240-22
JT9240-05CS 2141.39 USD
JT9240-05 CAJT9240-22 JT9240-7 CAJT9240-R JT9240-3 CAJT9240-3 JT9240-R CAJT9240-5 JT9240-22 CAJT9240-7
Diethyl ether ≥98% (by GC) stabilized, BAKER ANALYZED® ACS, J.T.Baker®
Diethyl ether

Stabilized with ethanol (1.5 to 3.5%), water (0.5%), and BHT (7 ppm). Lot analysis on label.

Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://us.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay ((C₂H₅)₂O) (by GC, exclusive of preservative) ≥98.0 %
Carbonyl Compounds (as HCHO) (by polarography) ≤0.001 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Titrable Acid (µeq/g) ≤0.2
Peroxide (as H₂O₂) ≤1 ppm
Preservative (C₂H₅OH), For Information Only
Preservative (H₂O), (w/w) ≤0.5 %
Preservative (BHT) ≥0.0007 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR