TCIAD1429

D1429-10G
TCD1429-010GEA 151.69 USD
TCD1429-010G
TCIAD1429
CAS Number: 725-12-2
MDL Number: MFCD00003102
Molecular Formula: C14H10N2O
Molecular Weight: 222.25
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Boiling point (°C): 231
Melting point (°C): 139
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR