1-Dodecanethiol ≥95.0%

Supplier: TCI America

Warning

Synonyms: 1-Mercaptododecane n-Dodecanethiol n-Dodecyl mercaptan

D0970-25ML D0970-500ML
TCD0970-25MLEA 15.94 USD
TCD0970-25ML TCD0970-500ML
1-Dodecanethiol ≥95.0%
1-Dodecanethiol

CAS Number: 112-55-0
MDL Number: MFCD00004885
Molecular Formula: C12H26S
Molecular Weight: 202.40
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 277
Melting point (°C): -7
Flash Point (°C): 128
Specific Gravity (20/20): 0.85
Lambda max.: 205 nm


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Dodecyl Mercaptan, Lauryl Mercaptan

Formula: C₁₂H₂₆S
MW: 202.4 g/mol
Boiling Pt: 277 °C
Melting Pt: –7 °C
Density: 0.85 (20 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00004885
CAS Number: 112-55-0
EINECS: 203-984-1
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR