2,6-Dichlorophenolindophenol sodium salt, powder, AR® ACS, Macron Fine Chemicals™

Synonyms: 2,6-Dichloroindophenol sodium derivativeTillmans' reagent2,6-Dichloroindophenol sodium saltSodium 4-(3,5-dichloro-4-oxocyclohexa-2,5-dienylideneamino)phenoxide2,6-Dichloroindophenol sodium

Management of Change (MOC) category = N

MACR3054-54
MK305454EA 520.41 USD
MK305454
2,6-Dichlorophenolindophenol sodium salt, powder, AR® ACS, Macron Fine Chemicals™
2,6-Dichlorophenolindophenol sodium salt
Formula: C₁₂H₆Cl₂NNaO₂
MW: 290.08 g/mol
Storage Temperature: Ambient
CAS Number: 620-45-1
EINECS: 210-640-4
Merck Index: 13,03093

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Loss on Drying at 120°C ≤12.0 %
ACS - Interfering Dyes Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR