1,5-Diphenylcarbazide, powder, BAKER ANALYZED® ACS, J.T.Baker®

Warning

Synonyms: sym. Diphenylcarbazidesym-DiphenylcarbazideN'',N'''-Diphenylcarbonohydrazide1,5-DiphenylcarbohydrazideDiphenylcarbazide1,5-Diphenylcarbonohydrazide

Management of Change (MOC) category = N

BAKRK620-03 BAKRK620-03 K620-05 BAKRK620-05
JTK620-5CS 1494.35 USD
CAJTK620-3 JTK620-5 JTK620-3 CAJTK620-5
1,5-Diphenylcarbazide, powder, BAKER ANALYZED® ACS, J.T.Baker®
1,5-Diphenylcarbazide

Powder.

Formula: C₁₃H₁₄N₄O
MW: 242.28 g/mol
Boiling Pt: 454 °C (1013 hPa)
Melting Pt: 170…172 °C
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00003013
CAS Number: 140-22-7
EINECS: 205-403-7
Merck Index: 13,03353
https://us.vwr.com/stibo/web/std.lang.all/46/13/2374613.jpg

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Melting Point 173 - 176 °C
Residue after Ignition ≤0.050 %
Solubility in Aqueous Acetone Passes Test
Sensitivity for Chromate Passes Test
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR