Diatomaceous earth filter aid, J.T.Baker®
|
Warning
|
Synonyms:
Celite®
, Diatomite
, Infusorial earth
, Diatomaceous silica
, Celite
, Kieselguhr
Management of Change (MOC) category = N
BAKR1939-05
BAKR1939-01
1939-01
BAKR1939-05
JT1939-1CS
524.4
USD
JT1939-1
CAJT1939-1
JT1939-5
CAJT1939-5
Diatomaceous earth filter aid, J.T.Baker®
Diatomaceous earth
Analytical filter aid.
Formula:
SiO₂ MW: 60.08 g/mol Boiling Pt: 2230 °C (1013 hPa) Melting Pt: 1710 °C Density: 1.9…2.35 g/cm³ (20 °C) |
MDL Number:
MFCD00132803 CAS Number: 68855-54-9 EINECS: 272-489-0 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Appearance | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...