Diethylamine 20% in acetonitrile for biosynthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
NC0017-0505
NC0017-0505
EM-NC0017-0505CS
3150
USD
CA-NC0017-0505
EM-NC0017-0505
Diethylamine 20% in acetonitrile for biosynthesis, Sigma-Aldrich®
Diethylamine
DEA/AcN Mix (20% DEA in AcN)
Formula:
(C₂H₅)₂NH MW: 73.14 g/mol Boiling Pt: 55.5 °C (1013 hPa) Melting Pt: –50 °C Density: 0.7048 g/cm³ (20 °C) Flash Pt: –39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009032 CAS Number: 109-89-7 EINECS: 203-716-3 UN: 1154 ADR: 3,II Merck Index: 13,03138 |
Specification Test Results
Assay (acetonitrile) | 79.5-80.5% (v/v) |
Assay (diethylamine) | 19.5-20.5% (v/v) |
Color (APHA) | 10 max |
Component: Diethylamine Purity | 98.0% min |
Form | Clear liquid |
Identity (IR-spectrum) | Conforms |
Water (H2O) | 150 ppm max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...