Diethylamine ≥98%, Supelco®
Supplier: MilliporeSigma
|
Danger
|
DX1045-5
DX1045-5
EM-DX1045-5EA
261
USD
EM-DX1045-5
CADX1045-5
Diethylamine ≥98%, Supelco®
Diethylamine
Formula:
(C₂H₅)₂NH MW: 73.14 g/mol Boiling Pt: 55.5 °C (1013 hPa) Melting Pt: –50 °C Density: 0.7048 g/cm³ (20 °C) Flash Pt: –39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009032 CAS Number: 109-89-7 EINECS: 203-716-3 UN: 1154 ADR: 3,II Merck Index: 13,03138 |
Specification Test Results
Assay | 98% min |
Color (APHA) | 30 max |
Form | Clear liquid |
Water (H2O) | 500ppm max |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...