1,2-Dichloroethane ≥99.0%, OmniSolv®, Supelco®

Danger

Synonyms: Ethylene dichlorideDichloroethaneEthylene chloride

DX0796-6 DX0796P-1 DX0796P-1 DX0796-6
EM-DX0796-6CS 2585.84 USD
CADX0796P-1 EM-DX0796-6 CADX0796-6 EM-DX0796P-1
1,2-Dichloroethane ≥99.0%, OmniSolv®, Supelco®
1,2-Dichloroethane

For spectrophotometry, chromatography, and residue analysis. UV cutoff 228nm. Filtered through a 0.2µm filter. Lot analysis on label.

Formula: ClH₂CCH₂Cl
MW: 98.96 g/mol
Boiling Pt: 83.5 °C (1013 hPa)
Melting Pt: –35.5 °C
Density: 1.254 g/cm³ (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00000963
CAS Number: 107-06-2
EINECS: 203-458-1
UN: 1184
ADR: 3,II
Merck Index: 13,03831
https://us.vwr.com/stibo/web/std.lang.all/54/88/725488.jpg

Order Now

Specification Test Results

Assay (GC) 99.0% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 500ppt max
Form Clear liquid
Free halogens To pass test
Infrared spectrum Conforms
Residue after evaporation 1ppm max
Titrable acid 0.3µeq/g max
UV Abs. at 230 nm 1.00AU max
UV Abs. at 240 nm 0.30AU max
UV Abs. at 250 nm 0.10AU max
UV Abs. at 300 nm 0.005AU max
UV Abs. at 400 nm 0.005AU max
UV Cut-off 230nm max
Water (H2O) 0.02% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR