Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CleaN-DEX™ 100% Nitrile Gloves, Best® Manufacturing

Supplier: Showa-Best Glove
CleaN-DEX™
C9905PFS C9905PFXL C9905PFM C9905PFL
ISO Class 5 (FED-STD-209E Class 100/M3.5) 100% nitrile gloves are suitable for critical environment applications, including electronic manufacturing and assembly, pharmaceuticals, biotechnology, and food processing

Ambidextrous, powder-free gloves provide a superior level of comfort and sensitivity. Rolled cuff, smooth finish. Easy don and removal process. Class II medical device. Gloves constructed according to materials and levels outlined in 12 CFR, 170-199. Manufactured in an ISO-9002 facility.

Particulate, extractables, and ESD levels fall well within electronics industry standards. Resistance level suitable for intermittant chemical handling and pharmaceutical uses.



 

ORDER

Expand All / Collapse All
Glove Size Length Thickness Glove Color Supplier No. VWR Catalog Number Unit Price Quantity
Large 28 cm (11") 5 mil White C9905PFL 61200-228 Case of 1000 Retrieving
Pack of 100 Retrieving
Medium 28 cm (11") 5 mil White C9905PFM 61200-226 Case of 1000 Retrieving
Pack of 100 Retrieving
Small 28 cm (11") 5 mil White C9905PFS 61200-224 Case of 1000 Retrieving
Pack of 100 Retrieving
X-Large 28 cm (11") 5 mil White C9905PFXL 61200-230 Case of 1000 Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Ambidextrous/Hand-specific Ambidextrous
Powdered/Powder-free Powder-Free

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty