N,N-Dimethylformamide, anhydrous ≥99.9% for applications requiring low water content, Burdick & Jackson™

Danger

Synonyms: NSC 5356Formic acid dimethylamideDimethylformamideDMFADMFFormyldimethylamineN-Formyldimethylamine

AS076-1 AS076-0100
CABJAS076-0100 CABJAS076-1
N,N-Dimethylformamide, anhydrous ≥99.9% for applications requiring low water content, Burdick & Jackson™
N,N-Dimethylformamide

Septum-sealed bottle


  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label

Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C
Melting Pt: -60.4 °C
Density: 0.949 g/cm3
Flash Pt: 58 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://us.vwr.com/stibo/web/std.lang.all/25/06/2302506.jpg https://us.vwr.com/stibo/web/std.lang.all/06/12/16640612.jpg

Order Now

Specification Test Results

Appearance Passes test
Color (APHA) 10 max.
GC Analysis 99.9 % min.
Residue 5 mg/L max.
Water by Karl Fischer Titration 50 ppm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR