1-Chloropropene ≥90.0%

Supplier: TCI America

Synonyms: 1-Chloro-1-propene1-Propenyl chloride

C0272-10ML
TCC0272-010MLEA 307.45 USD
TCC0272-010ML
1-Chloropropene ≥90.0%
1-Chloropropene

CAS Number: 590-21-6
MDL Number: MFCD00039358
Molecular Formula: C3H5Cl
Molecular Weight: 76.52
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 38
Flash Point (°C): -6
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: 1-Propenyl Chloride

Formula: C₃H₅Cl
MW: 76.53 g/mol
Boiling Pt: 38 °C
Melting Pt: –125 °C
Density: 0.92 (20 °C)
Flash Pt: –6 °C
MDL Number: MFCD00039358
CAS Number: 590-21-6
EINECS: 209-675-8

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR