Marble, chips, Macron Fine Chemicals™
Management of Change (MOC) category = N
MACR6210-06
MK621006CS
878.37
USD
MK621006
Marble, chips, Macron Fine Chemicals™
Marble
Large pieces or small chips of crystalline fracture from white marble slabs.
Formula:
CaCO₃ Melting Pt: 825 °C Density: 2.71 g/cm³ (20 °C) Storage Temperature: Ambient |
CAS Number:
471-34-1 EINECS: 207-439-9 Merck Index: 13,01658 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Identification | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...