Collodion USP, Macron Fine Chemicals™

Danger


Management of Change (MOC) category = R

MACR4560-08 MACR4560-04
MK456004CS 1536.23 USD
MK456004 MK456008
Collodion USP, Macron Fine Chemicals™
Collodion

Appearance: Clear or opalescent, colorless to slightly yellowish syrupy liquid which may contain some flocculent residue

Boiling Pt: ∼34 °C (1013 hPa)
Melting Pt: 160 °C
Density: 0.75 g/cm³ (20 °C)
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00081525
CAS Number: 9004-70-0
UN: 1993
ADR: 3,II
Merck Index: 12,08195

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (pyroxylin) ≥5.0 %
USP - Acidity Passes Test
USP - Alcohol Content 22.0 - 26.0 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Gravity at 25°/25°C 0.765 - 0.775
Appearance (clear or opalescent, colorless to slightly yellowishsyrupy
liquid which may contain some flocculent residue)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR