Charcoal activated 90.0-100.0%, powder USP, Macron Fine Chemicals™

Synonyms: Activated carbonCarbon activatedActivated charcoal

Management of Change (MOC) category = R

MACR4394-12
MK439412CS 959.28 USD
MK439412
Charcoal activated 90.0-100.0%, powder USP, Macron Fine Chemicals™
Charcoal activated

Activated powder.

Formula: C
MW: 12.01 g/mol
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specification Test Results

Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Acid-Soluble Substances ≤3.5 %
USP - Adsorptive power Passes Test
USP - Chloride (Cl) ≤0.2 %
USP - Cyanogen Compounds Passes Test
USP - Loss on Drying ≤15.0 %
USP - Microbial Limit Passes Test
USP - Reaction Passes Test
USP - Residue on Ignition ≤4.0 %
USP - Sulfate (SO₄) ≤0.2 %
USP - Sulfide (S) Passes Test
USP - Uncarbonized Constituents Passes Test
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR