Charcoal activated, powder, steam activated and acid washed, J.T.Baker®

Warning

Synonyms: Activated carbonCarbon activatedActivated charcoal

Management of Change (MOC) category = N

BAKRE345-07 E345-07
JTE345-7EA 179.22 USD
CAJTE345-7 JTE345-7
Charcoal activated, powder, steam activated and acid washed, J.T.Baker®
Charcoal activated

(NORIT® SX 2, formerly NORIT® SG EXTRA)


NORIT® - Registered Trademark of American Norit Company, Inc. ANALYTICAL

Formula: C
MW: 12.01 g/mol
Melting Pt: 3550 °C
Density: 2 g/cm³ (20 °C)
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
REACH: 01-2119488894-16
Merck Index: 13,01818

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
NORIT® - Registered Trademark of American Norit Company, Inc. ANALYTICAL
TEST THIS PRODUCT DOES NOT HAVE PRODUCT SPECIFICATION PARAMETERS. THE
TEST
DESCRIPTION, SPECIFICATION, AND RESULTS ARE INTENTIONALLY LEFT BLANK.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR