Collodion, flexible USP, J.T.Baker®

Danger


Management of Change (MOC) category = R

BAKR9204-01 9204-01
JT9204-1CS 1375.85 USD
JT9204-1 CAJT9204-1
Collodion, flexible USP, J.T.Baker®
Collodion

Flexible.

Boiling Pt: ∼34 °C (1013 hPa)
Melting Pt: 160 °C
Density: 0.75 g/cm³ (20 °C)
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00081525
CAS Number: 9004-70-0
UN: 2059
ADR: 3,II
Merck Index: 12,08195

Order Now

Specification Test Results

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Alcohol Content 21.0 - 25.0 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Gravity at 25°/25°C 0.770 - 0.790
USP - Presence of Camphor Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
STORAGE: Preserve in tight containers, at a temperature not exceeding
30°C, remote from fire.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR