2-(Cyclohexylamino)ethanesulfonic acid (CHES) ≥99.0% (dried basis), Ultrapure Bioreagent, J.T.Baker®

Synonyms: N-Cyclohexyltaurine

BAKR4146-01 BAKR4146-01
JT4146-1CS 863.93 USD
JT4146-1 CAJT4146-1
2-(Cyclohexylamino)ethanesulfonic acid (CHES) ≥99.0% (dried basis), Ultrapure Bioreagent, J.T.Baker®
2-(Cyclohexylamino)ethanesulphonic acid (CHES)

DNase-, RNase-, and protease-free. Actual lot analysis reported on label.

Formula: C₈H₁₇NO₃S
MW: 207.29 g/mol
Melting Pt: 320 °C
Storage Temperature: Ambient
MDL Number: MFCD00003835
CAS Number: 103-47-9
EINECS: 203-115-6
Merck Index: 13,02063

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₈H₁₇NO₃S) (dried basis) ≥ 99.0 %
Appearance Passes Test
Insoluble Matter ≤ 0.01 %
ACS - Heavy Metals (as Pb) ≤ 5 ppm
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
Water (by KF, volumetric) ≤ 1.0 %
Product Information (not specifications): pKa at 20°C: 9.30

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR