CAPS (3-(cyclohexylamino)propanesulfonic acid) ≥98.0%, Ultrapure Bioreagent, J.T.Baker®

Synonyms: 3-Cyclohexylaminopropanesulphonic acid3-Cyclohexylaminopropanesulfonic acid

Management of Change (MOC) category = N

BAKR4118-04 BAKR4118-04
JT4118-4EA 953.81 USD
CAJT4118-4 JT4118-4
CAPS (3-(cyclohexylamino)propanesulfonic acid) ≥98.0%, Ultrapure Bioreagent, J.T.Baker®
CAPS (3-(Cyclohexylamino)propanesulphonic acid)

Actual lot analysis reported on label. DNase-, RNase-, and protease-free.

Formula: C₆H₁₁NH(CH₂)₃SO₃H
MW: 221.32 g/mol
Melting Pt: 324 °C
Storage Temperature: Ambient
MDL Number: MFCD00003837
CAS Number: 1135-40-6
EINECS: 214-492-1
Merck Index: 13,01773

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (C₉H₁₉NO₃S) ≥98.0 %
Appearance Passes Test
Insoluble Matter ≤0.5 %
ACS - Heavy Metals (as Pb) ≤5 ppm
DNase Activity (None Detected) Passes Test
RNase Activity Passes Test
Protease Activity Passes Test
Product Information (not specifications):
pKa at 20°C.............................10.40

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR