Copper(I) oxide ≥96.0%, BAKER ANALYZED®, J.T.Baker®

Warning

Synonyms: Cuprous oxideDicopper oxide

Management of Change (MOC) category = N

BAKR1878-01 1878-01
JT1878-1CS 1153.8 USD
CAJT1878-1 JT1878-1
Copper(I) oxide ≥96.0%, BAKER ANALYZED®, J.T.Baker®
Copper (I) oxide

Powder.

Formula: Cu₂O
MW: 143.09 g/mol
Boiling Pt: 1800 °C (1013 hPa)
Melting Pt: 1232 °C
Density: 5.7…6.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010974
CAS Number: 1317-39-1
EINECS: 215-270-7
UN: 3077
ADR: 9,III
Merck Index: 13,02694

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (Cu₂O) ≥95.0 %
Preservative (zinc stearate) 0.2 - 0.5 %
Insoluble in HNO₃ ≤0.5 %
Chloride (Cl) ≤0.5 %
Sulfate (SO₄) ≤0.05 %
Iron (Fe) ≤0.05 %
Free Copper (Cu), For Information Only

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR