Cobalt(II) nitrate hexahydrate 98.0-102.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®

Danger

Synonyms: Cobalt dinitrate hexahydrateCobaltous nitrate hexahydrate

Management of Change (MOC) category = N

BAKR1680-01
JT1680-1CS 1416.11 USD
JT1680-1
Cobalt(II) nitrate hexahydrate 98.0-102.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®
Cobalt (II) nitrate hexahydrate

  • Lot analysis on label

Formula: Co(NO₃)₂·6H₂O
MW: 291.03 g/mol
Melting Pt: 57 °C
Density: 1.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,II
Merck Index: 13,02469

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (Co(NO₃)₂·6H₂O) (by EDTA titrn) 98.0 - 102.0 %
Insoluble Matter ≤0.01 %
pH of 5% Solution at 25°C 3.0 - 6.0
Chloride (Cl) ≤0.002 %
Sulfate (SO₄) ≤0.005 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.002 %
Trace Impurities - Iron (Fe) ≤0.001 %
Lead (Pb) ≤0.002 %
Magnesium (Mg) ≤0.005 %
Nickel (Ni) ≤0.15 %
Potassium (K) ≤0.01 %
Sodium (Na) ≤0.05 %
Zinc (Zn) ≤0.01 %
Meets Reagent Specifications for testing USP/NF monographs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR