Cobalt(II) chloride hexahydrate 98.0-102.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®
|
Danger
|
Synonyms:
Cobalt dichloride hexahydrate
, Cobalt Chloride, 6-Hydrate
, Cobaltous chloride hexahydrate
, Cobalt Chloride, Hexahydrate
Management of Change (MOC) category = N
BAKR1670-05
BAKR1670-01
BAKR1670-04
JT1670-5CS
10418.94
USD
JT1670-5
JT1670-4
JT1670-1
Cobalt(II) chloride hexahydrate 98.0-102.0% (by EDTA titration), BAKER ANALYZED® ACS, J.T.Baker®
Cobalt (II) chloride hexahydrate
Crystals. Lot analysis on label.
Formula:
CoCl₂·6H₂O MW: 237.93 g/mol Boiling Pt: 110 °C (1013 hPa) Melting Pt: 56 °C Density: 1.92 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149652 CAS Number: 7791-13-1 EINECS: 231-589-4 UN: 3288 ADR: 6.1,III Merck Index: 13,02462 |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Assay (CoCl₂·6H₂O) (by EDTA titrn) | 98.0 - 102.0 % |
Insoluble Matter | ≤0.01 % |
Nitrate (NO₃) | ≤0.01 % |
Sulfate (SO₄) | ≤0.01 % |
Calcium (Ca) | ≤0.005 % |
Copper (Cu) | ≤0.002 % |
Iron (Fe) | ≤0.005 % |
Magnesium (Mg) | ≤0.005 % |
Nickel (Ni) | ≤0.1 % |
Potassium (K) | ≤0.01 % |
Sodium (Na) | ≤0.05 % |
Zinc (Zn) | ≤0.03 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...