Chromium(III) oxide, powder, J.T.Baker®

Synonyms: Chromic oxide

Management of Change (MOC) category = N

BAKR1616-05 1616-01 BAKR1616-01 1616-05
JT1616-5CS 3295.5 USD
CAJT1616-5 JT1616-5 JT1616-1 CAJT1616-1
Chromium(III) oxide, powder, J.T.Baker®
Chromium (III) oxide
Formula: Cr₂O₃
MW: 151.99 g/mol
Boiling Pt: 4000 °C (1013 hPa)
Melting Pt: 2435 °C
Density: 5.22 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9
Merck Index: 12,02283

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ammonium (NH₄) ≤0.01 %
Chloride (Cl) ≤0.02 %
Iron (Fe) ≤0.05 %
Sodium (Na) ≤0.1 %
Sulfate (SO₄) ≤0.7 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR