Charcoal activated 90.0-100.0%, powder USP, J.T.Baker®
Synonyms:
Activated carbon
, Carbon activated
, Activated charcoal
Management of Change (MOC) category = R
BAKR1560-01
1560-01
JT1560-1CS
1337.49
USD
CAJT1560-1
JT1560-1
Charcoal activated 90.0-100.0%, powder USP, J.T.Baker®
Charcoal activated
Activated powder.
Formula:
C MW: 12.01 g/mol Melting Pt: 3550 °C Density: 2 g/cm³ (20 °C) |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 EINECS: 231-153-3 UN: 1362 ADR: 4.2,III REACH: 01-2119488894-16 Merck Index: 13,01818 |
Specification Test Results
Meets U.S.P Requirements | |
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
Microbial Limits - Escherichia coli | Passes Test |
Microbial Limits - Salmonella | Passes Test |
Reaction | Passes Test |
USP - Loss on Drying | ≤15.0 % |
Residue on Ignition | ≤4.0 % |
Acid-Soluble Substances | ≤3.5 % |
Chloride (Cl) | ≤0.2 % |
USP - Sulfate (SO₄) | ≤0.2 % |
USP - Sulfide (S) | Passes Test |
USP - Cyanogen Compounds | Passes Test |
USP - Uncarbonized Constituents | Passes Test |
Adsorptive Power: Alkaloids | Passes Test |
Adsorptive Power: Dyes | Passes Test |
Typical Particle Size, % thru 150 micron | ≥97 % |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...