Calcium hydrogen orthophosphate, anhydrous ≥98.0%, powder, BAKER ANALYZED®, J.T.Baker®

Synonyms: Calcium phosphate dibasicAnhydrous Dibasic Calcium PhosphateCalcium hydrogen phosphateCalcium perphosphate, ammonifiedCalcium phosphate, dibasic

Management of Change (MOC) category = N

1430-09 1430-05 BAKR1430-05 1430-07
JT1430-5CS 2169.95 USD
CAJT1430-09 JT1430-5 CAJT1430-5 CAJT1430-7
Calcium hydrogen orthophosphate, anhydrous ≥98.0%, powder, BAKER ANALYZED®, J.T.Baker®
Calcium hydrogen orthophosphate
Formula: CaHPO₄
MW: 136.06 g/mol
Melting Pt: 370 °C
Density: 2.89 g/cm³ (20 °C)
MDL Number: MFCD00010909
CAS Number: 7757-93-9
EINECS: 231-826-1
Merck Index: 12,01739

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CaHPO₄) ≥98.0 %
Chloride (Cl) ≤0.25 %
Sulfate (SO₄) ≤0.05 %
Nitrate (NO₃) ≤0.005 %
Arsenic (As) ≤3 ppm
Heavy Metals (as Pb) ≤0.003 %
Iron (Fe) ≤0.005 %
Insoluble in HCl ≤0.09 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR